Learn More
Veratramine, TRC
Product Code. 30455204
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455204 100 mg -
30452359 50 mg 50mg
30450931 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30455204 Supplier Toronto Research Chemicals Supplier No. TRCV127500100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60-70-8
Molecular Formula C27 H39 N O2
Molecular Weight (g/mol) 409.6
Synonym 3-Piperidinol, 5-methyl-2-[(1S)-1-[(3S,6aR,11aS,11bR)-2,3,4,6,6a,11,11a,11b-octahydro-3-hydroxy-10,11b-dimethyl-1H-benzo[a]fluoren-9-yl]ethyl]-, (2S,3R,5S)-, Veratraman-3,23-diol, 14,15,16,17-tetradehydro-, (3β,23β)-, Veratramine (7CI,8CI), 1H-Benzo[a]fluorene, veratraman-3,23-diol deriv., (2S,3R,5S)-5-Methyl-2-[(1S)-1-[(3S,6aR,11aS,11bR)-2,3,4,6,6a,11,11a,11b-octahydro-3-hydroxy-10,11b-dimethyl-1H-benzo[a]fluoren-9-yl]ethyl]-3-piperidinol, NSC 17821, NSC 23880
IUPAC Name (2S,3R,5S)-2-[(1S)-1-[(3S,6aR,11aS,11bR)-3-hydroxy-10,11b-dimethyl-1,2,3,4,6,6a,11,11a-octahydrobenzo[a]fluoren-9-yl]ethyl]-5-methylpiperidin-3-ol
SMILES C[C@H]([C@@H]1NC[C@@H](C)C[C@H]1O)c2ccc3[C@@H]4CC=C5C[C@@H](O)CC[C@]5(C)[C@H]4Cc3c2C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C27H39NO2/c1-15-11-25(30)26(28-14-15)17(3)20-7-8-21-22-6-5-18-12-19(29)9-10-27(18,4)24(22)13-23(21)16(20)2/h5,7-8,15,17,19,22,24-26,28-30H,6,9-14H2,1-4H3/t15-,17-,19-,22-,24-,25+,26-,27-/m0/s1
Formula Weight 409.2981
Chemical Name or Material Veratramine