Learn More
Verapamil Ethyl Methanethiosulfonate, Bromide, TRC
Product Code. 30455011
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455011 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455011 Supplier Toronto Research Chemicals Supplier No. TRCV12510025MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Verapamil Ethyl Methanethiosulfonate','Bromide
CAS 353270-25-4
Molecular Formula C30 H45 N2 O6 S2 . Br
Quantity 25 mg
InChI Formula InChI=1S/C30H45N2O6S2.BrH/c1-23(2)30(22-31,25-11-13-27(36-5)29(21-25)38-7)15-9-16-32(3,18-19-39-40(8,33)34)17-14-24-10-12-26(35-4)28(20-24)37-6;/h10-13,20-21,23H,9,14-19H2,1-8H3;1H/q+1;/p-1
SMILES [Br-].COc1ccc(CC[N+](C)(CCCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)CCSS(=O)(=O)C)cc1OC
IUPAC Name [4-cyano-4-(3,4-dimethoxyphenyl)-5-methylhexyl]-[2-(3,4-dimethoxyphenyl)ethyl]-methyl-(2-methylsulfonylsulfanylethyl)azanium;bromide
Molecular Weight (g/mol) 673.72
Formula Weight 672.1902