Learn More
Verapamil-d6 Hydrochloride, TRC
Product Code. 30456477
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456477 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30456477 Supplier Toronto Research Chemicals Supplier No. TRCV12500210MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Verapamil-d6 Hydrochloride
CAS 1185032-80-7
Purity Grade Notes HPLC
Molecular Formula C27 2H6 H32 N2 O4 . Cl H
Quantity 10 mg
Synonym α-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-(1-methylethyl)-benzeneacetonitrile-d6 Hydrochloride, Berkatens-d6, Calan-d6, Cardibeltin-d6, Dignover-d6, Isoptin-d6, Veracim-d6, Veramex-d6, Zolvera-d6
InChI Formula InChI=1S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20H,8,13-16H2,1-7H3;1H/i1D3,2D3;
SMILES Cl.[2H]C([2H])([2H])C(C([2H])([2H])[2H])C(CCCN(C)CCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-(1,1,1,3,3,3-hexadeuteriopropan-2-yl)pentanenitrile;hydrochloride
Molecular Weight (g/mol) 497.1
Formula Weight 496.2975
Percent Purity >95