missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VEGF Receptor 1 Monoclonal Antibody (5A4D2), Invitrogen™

Mouse Monoclonal Antibody
110.00 EUR - 361.00 EUR
Specifications
| Antigen | VEGF Receptor 1 |
|---|---|
| Clone | 5A4D2 |
| Concentration | 100 μg/mL |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|
30019297
|
Invitrogen™
MA552088 |
20 μL |
110.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019804
![]() |
Invitrogen™
MA552087 |
100 μL |
361.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
VEGF Receptor 1 Monoclonal antibody specifically detects VEGF Receptor 1 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifica
| VEGF Receptor 1 | |
| 100 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 10% trehalose, 0.02% Tween 80 and no preservative | |
| AI323757; embryonic receptor kinase 2; Emrk2; FLT; FLT1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; i21; sFlt1; soluble VEGFR 1; sVEGFR 1; Tyrosine-protein kinase FRT; tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGF R1; VEGFR1; VEGFR-1 | |
| Recombinant Human VEGFR1/FLT-1 protein (Ser 27-Asn756). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| FLT1 |
| 5A4D2 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P17948 | |
| 2321 | |
| IgG2a | |
| Protein A | |
| Antibody |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto