missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VEGF Receptor 1 Monoclonal Antibody (5A4D2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA552088
This item is not returnable.
View return policy
Description
FLT1 (VEGFR1) is a receptor tyrosine kinase which plays a critical role in angiogenesis. Active angiogenesis has been found to play important role in many pathological and physiological processes, such as tumor progression, diabetic retinopathy, rheumatoid arthritis and wound healing. May play an essential role as a negative regulator of embryonic angiogenesis by inhibiting excessive proliferation of endothelial cells. Can promote endothelial cell proliferation, survival and angiogenesis in adulthood. Its function in promoting cell proliferation seems to be cell-type specific. Promotes PGF-mediated proliferation of endothelial cells, proliferation of some types of cancer cells, but does not promote proliferation of normal fibroblasts (in vitro). Has very high affinity for VEGFA and relatively low protein kinase activity; may function as a negative regulator of VEGFA signaling by limiting the amount of free VEGFA and preventing its binding to KDR.
Specifications
| VEGF Receptor 1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 10% trehalose, 0.02% Tween 80 and no preservative | |
| P17948 | |
| FLT1 | |
| Recombinant Human VEGFR1/FLT-1 protein (Ser 27-Asn756). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Immunocytochemistry | |
| 5A4D2 | |
| Unconjugated | |
| FLT1 | |
| AI323757; embryonic receptor kinase 2; Emrk2; FLT; FLT1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; i21; sFlt1; soluble VEGFR 1; sVEGFR 1; Tyrosine-protein kinase FRT; tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGF R1; VEGFR1; VEGFR-1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2321 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction