missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ VDR Recombinant Human Protein, His Tag
Description
Expressed from DNA sequence encoding human VDR (Met 1-Ser 427) fused to polyhistidine tag at C-terminus
- Formulation: 50mM Tris, 100mM NaCl, 10% Glycerol, pH 8.0
Specifications
Specifications
| For Use With (Application) | Protein Structure Analysis, Assay Standard |
| Gene ID (Entrez) | 7421 |
| Molecular Weight (g/mol) | 50kd |
| Quantity | 5 x 5 μg |
| Source | Human |
| Structural Form | Full Length, Recombinant |
| Endotoxin Concentration | <1 EU/μg |
| Common Name | Vitamin D(1,25- Dihydroxyvitamin D3)Receptor |
| Gene Symbol | VDR |
| Product Type | VDR Recombinant Human Protein |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?