Learn More
Eppendorf™ Varitip S Tips
Product Code. 10363481 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
2.5 to 10 mL
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10363481 2.5 to 10 mL Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10363481 Supplier Eppendorf™ Supplier No. 0030050568
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
Bulk Quote Request
Use these tips to meet a variety of needs for different lab applications. Eppendorf™ Varitip S Tips form a system with the Varitip and are used for aspirating liquid from tall, narrow-necked vessels.
  • A continuously adjustable pipettor for precise pipetting of volumes up to 10mL in 10μL increments
  • Even liquids with a high vapor pressure or viscosity can be pipetted with ease
  • System consists of a dispensing part and the Maxitip air displacement principle
  • No carry over as Maxitip exchanged
  • Valve for Maxitip ensures drip free dispensing of liquids with a high vapor pressure
  • Graduated Maxitip G for compensation of nominal value of Varipette™
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 2.5 to 10 mL
For Use With (Application) Used for aspirating liquid eppendorf
Graduated Yes