Learn More
MedChemExpress Vapreotide
Specifications
Specifications
| Purity | 98.75% |
| CAS | 103222-11-3 |
| Content And Storage | Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 1131.37 g/mol |
| Product Type | Peptides |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 100 mg/mL (88.39 mM∣Need ultrasonic) |
| Synonym | RC160∣ BMY 41606 |