missing translation for 'onlineSavingsMsg'
Learn More

Vanillin oxime, 98%, Thermo Scientific™

Product Code. 15439099
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439099 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15439099 Supplier Thermo Scientific Alfa Aesar Supplier No. L19667.06
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2874-33-1
Melting Point 118°C to 122°C
Molecular Formula C8H9NO3
MDL Number MFCD00019965
Quantity 5 g
Beilstein 1952864
Synonym 4-hydroxy-3-methoxybenzaldehyde oxime, 3-methoxy-4-hydroxy benzaldehyde oxime, vanillin oxime, 4e-4-hydroxyamino methylidene-2-methoxycyclohexa-2,5-dien-1-one, benzaldehyde, 4-hydroxy-3-methoxy-, oxime, 3-methoxy-4-hydroxybenzaldehyde oxime, 4-e-hydroxyimino methyl-2-methoxyphenol, 4e-2-methoxy-4-oxidanylamino methylidene cyclohexa-2,5-dien-1-one, 4e-4-hydroxyamino methylidene-2-methoxy-1-cyclohexa-2,5-dienone
InChI Key WKYQAOKXBUWYNX-AATRIKPKSA-N
SMILES COC1=C\C(=C\NO)C=CC1=O
IUPAC Name (4E)-4-[(hydroxyamino)methylidene]-2-methoxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol) 167.16
PubChem CID 5484330
Formula Weight 167.17
Percent Purity 98%
Chemical Name or Material Vanillin oxime
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.