missing translation for 'onlineSavingsMsg'
Learn More

Valsartan N1-Glucuronide, TRC

Product Code. 30456035
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456035 5 mg 5mg
30450609 25 mg 25mg
30449214 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456035 Supplier Toronto Research Chemicals Supplier No. TRCV0957905MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valsartan N1-Glucuronide
Analyte or Component Names Valsartan N1-Glucuronide
Shipping Condition Room Temperature
Molecular Formula C30 H37 N5 O9
InChI Formula InChI=1S/C30H37N5O9/c1-4-5-10-21(36)34(22(16(2)3)29(40)41)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)27-31-32-33-35(27)28-25(39)23(37)24(38)26(44-28)30(42)43/h6-9,11-14,16,22-26,28,37-39H,4-5,10,15H2,1-3H3,(H,40,41)(H,42,43)/t22-,23-,24-,25+,26-,28+/m0/s1
Solution Type Neat
SMILES CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnnn3[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O)[C@@H](C(C)C)C(=O)O
IUPAC Name (2S,3S,4S,5R,6R)-6-[5-[2-[4-[[[(1S)-1-carboxy-2-methylpropyl]-pentanoylamino]methyl]phenyl]phenyl]tetrazol-1-yl]-3,4,5-trihydroxyoxane-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 611.643
Quantity 5 mg
Formula Weight 611.259
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.