missing translation for 'onlineSavingsMsg'
Learn More

Valinomycin, ≥93%, MP Biomedicals™

Product Code. 11481812 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481812 5 mg 5mg
11491812 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481812 Supplier MP Biomedicals Supplier No. 0210501005
  • ONLINE EXCLUSIVE PRICE
    40.80 EUR
    38.85 EUR / 5mg
    Save 1.95 EUR
    4% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2001-95-8
Molecular Formula C54H90N6O18
Molecular Weight (g/mol) 1111.34
MDL Number MFCD00005114
InChI Key FCFNRCROJUBPLU-UYBNATROSA-N
PubChem CID 131668508
IUPAC Name (3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
SMILES CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C

Specifications

Type Valinomycin
Physical Form Crystalline Powder
Color White
Quantity 5 mg
Assay Percent Range ≥93%
Formula Weight 1111.4
Grade Not Available
Chemical Name or Material Valinomycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.