Learn More
Welch Ilmvac™ Vacuum Filtration And Desiccation Connection Kit
Product Code. 13504920 Shop All Welch Ilmvac Products
Change view
Click to view available options
Tubing Size:
6 mm ID, 8 mm ID
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Unit Size
13504920 6 mm ID, 8 mm ID Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13504920 Supplier Welch Ilmvac™ Supplier No. 404008
  • ONLINE EXCLUSIVE PRICE
    110.00 EUR
    85.30 EUR / Pack of 1
    Save 24.70 EUR
    22% Off
Estimated Shipment: 02-12-2026
Facilitates proper connection to vacuum filtration or dissicator. Welch Ilmvac™ Vacuum Filtration And Desiccation Connection Kit includes a filter and suitable tubing.
  • Quick connection to vacuum filtration funnel / manifold or dessicator
  • Kit includes in-line hydrophobic filter, 1.5m silicone vacuum hose (6mm I.D.) and 1.5m silicone vacuum hose (8mm I.D.)
TRUSTED_SUSTAINABILITY
Specifications
Includes In-Line Hydrophobic Filter, 1.5m Silicone Vacuum Hose (6mm ID) And 1.5m Silicone Vacuum Hose (8mm ID)
Quantity 1 Kit
Tubing Size 6 mm ID, 8 mm ID
Type Vacuum Filtration And Desiccation Connection Kit