Learn More
BRANDTECH™ VACUUBRAND™ Electronic Vacuum Gauges
Description
- DVR2pro model is battery-powered for portability, with a capacitive transducer for absolute pressure measurements from atmosphere to 1 mbar/Torr/hPA. Combination digital/analog scale for easy reading.
- VACUU VIEW model features a capacitive transducer for absolute pressure measurement and measures to 0.1 mbar/Torr/hPA. Universal power suppy.
- VACUU VIEW extended, has a combination capacitive/Pirani transducer for measurements to 10-3 mbar/Torr/hPA. Universal power supply.
Specifications
Specifications
| Display | Digital |
| For Use With (Application) | Vacuum applications in the rough/coarse vacuum range atmosphere - 0.1 mbarTorr/hPa |
| For Use With | Vacuum applications |
| Includes | Vacuum gauge, power supply, KF16 small flange connection and screw in 6/10mm hose barb |
| Material | Various |
| Product Line | VACUU VIEW |
| Type | Capacitive Vacuum Gauge |
| Weight (Metric) | 1 kg |
| Height (Metric) | 8.5 cm |
| Length (Metric) | 25 cm |
| Show More |