Learn More
BRANDTECH™ VACUUBRAND™ Electronic Vacuum Gauges
Product Code. 15509311
Change view
Click to view available options
Maximum Pressure:
0.001 to 1100 mbar
0.1 to 1100 mbar
1 to 1060 mbar
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Maximum Pressure Unit Size
15509311 0.001 to 1100 mbar Each
15838894 0.1 to 1100 mbar Each
15711368 1 to 1060 mbar Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15509311 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20683210
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 07-12-2026
VACUUBRAND vacuum gauges are mercury-free. Choose from models for the coarse/rough vacuum range or fine vacuum range. User-selectable units.
  • DVR2pro model is battery-powered for portability, with a capacitive transducer for absolute pressure measurements from atmosphere to 1 mbar/Torr/hPA. Combination digital/analog scale for easy reading.
  • VACUU VIEW model features a capacitive transducer for absolute pressure measurement and measures to 0.1 mbar/Torr/hPA. Universal power suppy.
  • VACUU VIEW extended, has a combination capacitive/Pirani transducer for measurements to 10-3 mbar/Torr/hPA. Universal power supply.
TRUSTED_SUSTAINABILITY
Specifications
Display Digital
For Use With (Application) Vacuum applications in the rough and fine vacuum ranges atmosphere to 1x10-3 mbar/Torr/hPa
For Use With Vacuum pumps
Includes Vacuum gauge, power supply, KF16 flange connection and 6/10mm screw in hose barb
Material Various
Product Line VACUU VIEW extended
Type Capacitive/Pirani Vacuum Gauge
Weight (Metric) 1 kg
Height (Metric) 8.5 cm
Length (Metric) 25 cm
Width (Metric) 16 cm
Maximum Pressure 0.001 to 1100 mbar