Learn More
BD Vacutainer™ Push Button Blood Collection Sets
Product Code. 12947746
Click to view available options
Needle Gauge:
21 G
Length (Metric) Tube:
178 mm
Unit Size:
Pack of 100
Product Code. 12947746
Brand: BD 367355
Currently Unavailable: We have no information when this product will be available.
View alternative products
Specifications
Product Type Push Button Blood Collection Set
Color Green
Needle Gauge 21 G
Includes 21 guage, 19mm needle size, 178mm tubing
Length (Metric) Needle 19 mm
Length (Metric) Tube 178 mm
Quantity 100 Cs.