Learn More
Greiner Bio-One Vacuette Tube™, Premium, Red Cap-Yellow Ring, CAT Serum Separator Clot Activator, Sterile
Product Code. 10060704
Change view
Click to view available options
Capacity (Metric):
2.5 mL
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10060704 2.5 mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10060704 Supplier Greiner Bio-One Supplier No. 454028
  • ONLINE EXCLUSIVE PRICE
    18.32 EUR
    11.10 EUR / Pack of 50
    Save 7.22 EUR
    39% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Includes inner wall coated with microscopic silica particles to activate coagulation and a separator gel to separate serum and blood cells.
  • The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.
  • Serum separator tubes contain a separator gel in the base of the tube; during centrifugation, this gel forms a stable barrier between the serum and the blood cells.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vacuette Tube
Additive Clot Activator and Gel
Sterility Sterile
Capacity (Metric) 2.5 mL
Closure Color Red with Yellow Ring
Length (Metric) 75 mm
Diameter (Metric) Outer 13 mm