Learn More
UVP Compact and Handheld UV Lamp Replacement Tubes
Product Code. 11902041
Change view
Click to view available options
Wavelength:
254 nm
365 nm
Wattage:
4 W
6 W
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Wattage Unit Size
11902041 254 nm 6 W Each
12812979 254 nm 4 W Each
11786832 365 nm 4 W Each
12862979 365 nm 6 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11902041 Supplier UVP Supplier No. 34001301
Estimated Shipment: 17-12-2026
For use with 4w and 6w UV lamps.
  • For use with 4w and 6w UV lamps
Specifications
Light Source UV-C
Voltage 115 V
Wattage 6 W
Wavelength 254 nm
For Use With (Application) For use with Compact 4-Watt UV Lamp
For Use With (Equipment) Compact 4-Watt UV Lamp
Product Type UV Lamp Tube