missing translation for 'onlineSavingsMsg'
Learn More

UVI 3003, MedChemExpress

Product Code. 16406847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16406847 5 mg 5mg
16494138 10 mg 10mg
16462966 25 mg 25mg
16420497 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16406847 Supplier Medchem Express Supplier No. HY107500
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress UVI 3003 is a highly selective antagonist of retinoid X receptor (RXR), and inhibits xenopus and human RXRα in Cos7 cells, with IC50s of 0.22 and 0.24 μM, respectively.

UVI 3003 is a highly selective antagonist of retinoid X receptor (RXR), and inhibits xenopus and human RXRα in Cos7 cells, with IC50s of 0.22 and 0.24 μM, respectively.

Specifications

Chemical Name or Material UVI 3003
CAS 847239-17-2
Color White
Purity Grade Notes Research
Molecular Formula C28H36O4
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (229.05 mM; Need ultrasonic)
SMILES CCCCCOC1=C(C2=CC(/C=C/C(O)=O)=CC=C2O)C=C3C(C(C)(CCC3(C)C)C)=C1
Molecular Weight (g/mol) 436.58
Formula Weight 436.58
Percent Purity 99.03%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.