Learn More
Uvex™ Wet Wipes
Product Code. 15786108 Shop All Uvex Products
Change view
Click to view available options
:
Product Type: Wet Wipes
Product Code.
15786108 Product Type: Wet Wipes
1 options
Product Code. 15786108
Brand: Uvex™ 9963000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Dimensions (L x W) 8 x 5 in.
Description Individually wrapped
Product Type Lens Cleaning Towelette
For Use With (Equipment) Polycarbonate, plastic, and glass lenses
For Use With (Application) Cleaning Safety Eyewear
Includes Dispenser box
No. per Pack 100