Learn More
Uvex™ Wet Wipes
Product Code. 15356272 Shop All Uvex Products
Change view
Click to view available options
Product Type: Wet Wipes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15356272 Product Type: Wet Wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15356272 Supplier Uvex™ Supplier No. 9963000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Dimensions (L x W) 8 x 5 in.
Description Individually wrapped
Product Type Lens Cleaning Towelette
For Use With (Equipment) Polycarbonate, plastic, and glass lenses
For Use With (Application) Cleaning Safety Eyewear
Includes Dispenser box
No. per Pack 100