Learn More
Jenway™ 72 Series UV/Visible Spectrophotometer
Product Code. 15641227
Change view
Click to view available options
Detector Type:
Photodiode Array
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detector Type Unit Size
15641227 Photodiode Array Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15641227 Supplier Jenway™ Supplier No. 8305602
  • ONLINE EXCLUSIVE PRICE
    3535.00 EUR
    3168.48 EUR / Pack of 1
    Save 366.52 EUR
    10% Off
View all promotions
Estimated Shipment: 02-12-2026
The first scanning spectrophotometers in the Jenway range to leverage diode array technology to produce exceptionally fast results.
  • Scanning diode array technology
  • Color touchscreen navigation
  • Small footprint and lightweight
  • Fast scan speed
  • English, French and German language options
  • Multiple USB ports for data storage and printer connectivity
  • Extensive range of accessories available
  • 2 year warranty (including xenon lamp)
  • Ideal for a variety of applications in education and routine testing in clinical, veterinary, pharmaceutical and QC laboratories
Specifications
Absorbance Accuracy ±0.01A at 1.0A and 546nm
Absorbance Range −0.300 to +2.500 A
Beam Center Height 15 mm
Concentration ±2500
Detector Type Photodiode Array
Dimensions (L x W x H) 212 x 422 x 120 mm
Spectral Bandwidth 5 nm
Voltage 100/240 V
Wavelength Range 198 to 800 nm
Interface Color touch screen
Noise ±0.002A at 0.1A and ±0.02A at 2.0A and 546nm
Optical Design Photodiode Array
Photometric Repeatability ±2 nm
Printer Included Optional
Stray Light At 340 nm: < 1%T according to ANSI/ASTM
Warranty 2 years including Xenon lamp
Wavelength Accuracy ±2 nm
Wavelength Repeatability ±2 nm
Weight (Metric) 2.8 kg
Frequency 50/60 Hz