Learn More
Fisherbrand™ UV Sterilization Cabinet with Timer
Product Code. 15562496 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
No tray
Tray
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
15562496 No tray Each
15592496 Tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15562496 Supplier Fisherbrand™ Supplier No. FUVCAB
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Limits PCR reaction contamination. Fisherbrand™ UV Sterilization Cabinet with Timer provides a convenient area for setting up PCR reactions in a nucleic acid-free environment.
  • UV lights on the cabinets denature nucleic acids in 5 to 30 minutes making them unsuitable for amplification.
  • Incorporates safety features to prevent user exposure to UV light.
  • The UV lights are timer controlled
  • Safety switches on the cabinets doors turn off the UV lights when opened.
  • Shields beta radioactive emissions
  • Specifications
    Lighting Type 4 UV Lights, 1 White
    Depth (Metric) 420 mm
    Height (Metric) 770 mm
    Includes No tray
    Wattage 5 x 15 W
    Weight (Metric) 19 kg
    Width (Metric) 560 mm