Learn More
Cole-Parmer™ UV-A/B Light Meter
Product Code. 12899782 Shop All Cole Parmer Products
Change view
Click to view available options
Wavelength:
280 to 400 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Unit Size
12899782 280 to 400 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12899782 Supplier Cole-Parmer™ Supplier No. 5051300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Measure damaging UV (ultraviolet) light
  • Conduct UV degradation studies for pharmaceutical, cosmetic, semiconductor, printing, forensic applications and more.
  • The UV light intensity appears on the large backlit display in either ìW or mW/cm² while the small lower display simultaneously shows minimum, maximum, average, and recorded data.
  • Up to 20 data points can be recorded automatically at a variety of intervals.
  • The UV-A/B model measures long and medium wave UV-A/B for forensics, chromatography, electrophoresis, and dermatology applications.
Specifications
Accuracy ±4% ±1 digit (at 23 ± 5°C)
Height (Metric) 15.2 cm
Length (English) 1 1/2 in.
Length (Metric) 3.8 cm
Resolution Intensity (mW/cm2):40
Wavelength 280 to 400 nm
Width (English) 2 in.
Width (Metric) 5.1 cm
Display Backlit LCD