Learn More
UV-320, TRC
Product Code. 30452508
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452508 25 g 25g
30456157 50 g 50g
30454269 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30452508 Supplier Toronto Research Chemicals Supplier No. TRCU85021025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3846-71-7
Molecular Formula C20 H25 N3 O
Molecular Weight (g/mol) 323.43
Synonym Phenol, 2-(2H-benzotriazol-2-yl)-4,6-di-tert-butyl- (7CI,8CI), 2-(2H-Benzotriazol-2-yl)-4,6-bis(1,1-dimethylethyl)phenol, 2-(2-Benzotriazolyl)-4,6-di-tert-butylphenol, 2-(2-Hydroxy-3,5-di-tert-butylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-3,5-di-tert-butylphenyl)benzotriazole, 2-(2'-Hydroxy-3',5'-di-t-butylphenyl)benzotriazole, 2-(2'-Hydroxy-3',5'-di-tert-butylphenyl)benzotriazole, 2-(2'-Hydroxy-3'5-di-tert-butylphenyl) benzotriazole, 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole, 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)benzotriazole, 2-(3',5'-Di-tert-butyl-2'-hydroxyphenyl)benzotriazole, Eversorb 77, Kemisorb 75, Seesorb 705, Sumisorb 320, Tinuvin 320, UV 320, Viosorb 582
IUPAC Name 2-(benzotriazol-2-yl)-4,6-ditert-butylphenol
SMILES CC(C)(C)c1cc(c(O)c(c1)C(C)(C)C)n2nc3ccccc3n2
Specifications
Quantity 25 g
InChI Formula InChI=1S/C20H25N3O/c1-19(2,3)13-11-14(20(4,5)6)18(24)17(12-13)23-21-15-9-7-8-10-16(15)22-23/h7-12,24H,1-6H3
Formula Weight 323.1998
Percent Purity >95%
Chemical Name or Material 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole
Grade HPLC