Learn More
MedChemExpress Uridine triphosphate
Specifications
Specifications
| Molecular Weight (g/mol) | 484.14 g/mol |
| Product Type | Natural Products |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 98.0% |
| Quantity | 10 mg |
| Solubility Information | H2O : ≥ 150 mg/mL (309.83 mM) |
| For Use With (Application) | Cancer-Kinase/protease |
| CAS | 63-39-8 |
| Synonym | UTP∣ Uridine 5'-triphosphate |