Learn More
Uridine-5'-triphosphate Trisodium Salt Dihydrate (>90%), TRC
Product Code. 30455367
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455367 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455367 Supplier Toronto Research Chemicals Supplier No. TRCU830048500MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Uridine-5'-triphosphate Trisodium Salt Dihydrate
CAS 116295-90-0
Molecular Formula C9 H12 N2 Na3 O15 P3 . 2(H2 O)
Quantity 500 mg
Synonym Uridine 5'-(tetrahydrogen triphosphate) sodium salt hydrate (1:3:2), Uridine 5'-(tetrahydrogen triphosphate) trisodium salt dihydrate (9CI), UTP trisodium dihydrate
InChI Formula InChI=1S/C9H15N2O15P3.3Na.2H2O/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;;;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H,10,12,15)(H2,16,17,18);;;;2*1H2/q;3*+1;;/p-3/t4-,6-,7-,8-;;;;;/m1...../s1
SMILES O.O.[Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)(O)OP(=O)([O-])OP(=O)([O-])[O-])N2C=CC(=NC2=O)O
IUPAC Name trisodium;[[[(2R,3S,4R,5R)-3,4-dihydroxy-5-(4-hydroxy-2-oxo-pyrimidin-1-yl)tetrahydrofuran-2-yl]methoxy-hydroxy-phosphoryl]oxy-oxido-phosphoryl] phosphate;dihydrate
Molecular Weight (g/mol) 586.12
Formula Weight 585.9355