missing translation for 'onlineSavingsMsg'
Learn More

Uridine-5'-Triphosphate Trisodium salt, 90 to 95%, MP Biomedicals™

Product Code. 11430292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
Unit Size:
100mg
1g
250mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430292 100 mg 100mg
11440292 250 mg 250mg
11450292 500 mg 500mg
11460292 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11430292 Supplier MP Biomedicals Supplier No. 0219123080
  • ONLINE EXCLUSIVE PRICE
    61.75 EUR
    58.80 EUR / 100mg
    Save 2.95 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 19817-92-6
Molecular Formula C9H12N2Na3O15P3
Molecular Weight (g/mol) 550.09
MDL Number MFCD11045990
InChI Key MMJGIWFJVDOPJF-ZKGVITBDNA-K
Synonym uridine-5'-triphosphate trisodium salt dihydrate, uridine 5'-triphosphate trisodium salt dihydrate, trisodium uridine 5'-triphosphate dihydrate, 550.0866 anhydrous, c9h12n2na3o15p3.xh2o, uridine 5'-tetrahydrogen triphosphate , sodium salt, hydrate 1:3:2, trisodium dihydrate utp trianion, sodium 5'-o-hydroxyphosphinato oxy phosphinato oxy phosphinato uridine hydrate 3:1:2, trisodium, 2r,3s,4r,5r-5-2,4-dioxopyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl hydrogen phosphate, dihydrate, uridine 5'-triphosphate, trisodium salt, hydrate
PubChem CID 131675549
IUPAC Name trisodium ({[(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(hydrogen phosphonatooxy)phosphinate
SMILES [Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N1C=CC(=O)NC1=O

Specifications

Physical Form Solid
Quantity 100 mg
Assay Percent Range 90 to 95%
Formula Weight 550.09
Appearance Powder
Chemical Name or Material Uridine-5'-Triphosphate Trisodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.