missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5’-Triphosphate Trisodium Hydrate, TRC

Product Code. 30453013
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453013 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453013 Supplier Toronto Research Chemicals Supplier No. TRCU830025250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Uridine 5'-Triphosphate Trisodium Salt Hydrate
CAS 19817-92-6
Molecular Formula C9H12N2Na3O15P3•xH2O
Quantity 250 mg
Synonym Uridine 5'-(Tetrahydrogen Triphosphate) Trisodium Hydrate, NSC 20260 Hydrate, Trisodium UTP Hydrate, UTP Trisodium Hydrate, Utipina Hydrate
SMILES O[C@@H]1[C@H](O)[C@@H](COP(OP(OP(O)(O[Na])=O)(O[Na])=O)(O[Na])=O)O[C@H]1N(C=C2)C(NC2=O)=O
Molecular Weight (g/mol) 550.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.