missing translation for 'onlineSavingsMsg'
Learn More

Uridine-5'-diphosphate disodium salt, 98%

Product Code. p-7042914
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496319 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15496319 Supplier Thermo Scientific Alfa Aesar Supplier No. J64329.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27821-45-0 | C9H12N2Na2O12P2 | 448.12 g/mol

Uridine-5'-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uridine-5′-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).

Solubility
Slightly soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27821-45-0
Molecular Formula C9H12N2Na2O12P2
Molecular Weight (g/mol) 448.12
MDL Number MFCD00006524,MFCD00084679,MFCD04975448
InChI Key ZQKVPFKBNNAXCE-HVQNCJTOSA-L
Synonym 5'-udp,2na
PubChem CID 131676107
SMILES [Na+].[Na+].OC1C(O)[C@@H](O[C@@H]1COP(O)(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O

Specifications

Color White
Quantity 1 g
Beilstein 8817400
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
IUPAC Name disodium;(2R,3R,4S,5R)-2-(2,4-dioxopyrimidin-1-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
Formula Weight 448.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Uridine-5'-diphosphate disodium salt
compliance-icons
Product Identifier
  • Uridine-5'-diphosphate disodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Uridine-5'-diphosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.