Learn More
Urethane-13C,15N, TRC
Specifications
Specifications
| Chemical Name or Material | Urethane-13C,15N |
| Molecular Formula | 13C C2 H7 15N O2 |
| Quantity | 10 mg |
| Synonym | Carbamic Acid Ethyl Ester-13C,15N, Ethyl Aminoformate-13C,15N, Ethyl Carbamate-13C,15N, Ethylurethane-13C,15N, Leucethane-13C,15N, NSC 746-13C,15N, O-Ethylurethane-13C,15N, Pracarbamine-13C,15N, Urethan-13C,15N |
| InChI Formula | InChI=1S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5)/i3+1,4+1 |
| SMILES | CCO[13C](=O)[15NH2] |
| IUPAC Name | ethyl carbamate |
| Molecular Weight (g/mol) | 91.079 |
| Formula Weight | 91.048 |