missing translation for 'onlineSavingsMsg'
Learn More

Urea, TRC

Product Code. 30007701
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007701 50 g 50g
30007036 100 g 100g
17977904 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007701 Supplier Toronto Research Chemicals Supplier No. TRCU82250050G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula C H4 N2 O
Molecular Weight (g/mol) 60.055
Synonym Urea, AUS 32, AdBlue, Aquacare, Aquadrate, B-I-K, Basodexan, Benural 70, Carbamide, Carbamimidic acid, Carbonyl diamide, Carmol 40, Cellpaste K 4, Cellton NP, DEF, DEF (diesel exhaust fluid), Diesel exhaust fluid, Duration III, ESN, Elaqua XX, Eucerin 10% Urea Lotion, Hyanit, Isourea, Keratinamin, Keratinamin Kowa, Low Biuret Urea, NSC 34375, Nutraplus, Onychomal, Onyster, Optigen 1200, Optigen II, Pastaron, Pastaron 10, Pastaron 20, Pastaron 20 soft, Pastaron soft, Pseudourea, Rubinol ST 010, SGN 250, UR, Ultra Low Biuret Urea, Urea perhydrate, Ureaphil, Ureophil, Urepeal, Urepeal L, Urepearl, Urevert, Uria, Uroderm, Varioform II
IUPAC Name urea
SMILES O=C(N)N

Specifications

Quantity 50 g
InChI Formula InChI=1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)
Formula Weight 60.032
Percent Purity >95%
Chemical Name or Material Urea
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.