missing translation for 'onlineSavingsMsg'
Learn More

Uranine AP (C.I. 45350), Supelco™

Product Code. 15815160
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15815160 1 kg 1kg
15805160 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15815160 Supplier Supelco Supplier No. 1084621000
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Concentrated for the examination of subterranean waters

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 518-47-8

Specifikationer

Melting Point >300°C
MDL Number MFCD00167039
Quantity 1 kg
Synonym Uranine AP (C.I. 45350), Uranine AP (C.I. 45350)
Molecular Weight (g/mol) 376.27
Grade Research
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.