Learn More
Uracil-d4, CDN
Specifications
Specifications
| Chemical Name or Material | Uracil (D4) |
| CAS | 24897-55-0 |
| Molecular Formula | C42H4N2O2 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2 H,(H2,5,6,7,8)/i1D,2 D/hD2 |
| SMILES | [2 H]N1C(=C([2 H])C(=O)N([2 H])C1=O)[2 H] |
| IUPAC Name | 1,3,5,6-tetradeuteriopyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 116.11 |
| Formula Weight | 116.0524 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |