Learn More
Fisherbrand™ Upright ULT Freezer Racks for Microtiter Plates
Product Code. 15749559 Shop All Fisherbrand Traceable Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Holds:
Plates with Lid (70)
Plates with Lid (84)
Plates with Lid (96)
missing translation for 'unitSize':
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds missing translation for 'unitSize'
15749559 Plates with Lid (96) Each
15729559 Plates with Lid (70) Each
15739559 Plates with Lid (84) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Product Code. 15749559
  • ONLINE EXCLUSIVE PRICE
    87.54 EUR
    73.69 EUR / Pack of 1
    Save 13.85 EUR
    15% Off
View all promotions
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Fisherbrand inventory control racks for 96-Well and 384-Well microtiter plates. They fit all standard types of upright ULT freezers.
  • Holds up to 96 plates with lid or 120 plates without lid
  • Made of high quality stainless steel
  • Side opening with handles on the front for easy plate retrieval.
Specifications
Holds Plates with Lid (96)
Material Stainless Steel
For Use With (Application) 96/384-Well Microtiter Plates
For Use With (Equipment) Upright ULT Freezer
Dimensions (L x W x H) 559 x 280 x140 mm
Product Type Side Access Freezer Rack