Learn More
Fisherbrand™ Upright ULT Freezer Racks
Product Code. 15739539 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
12 (3in.) boxes
12 Boxes (2 in.)
15 (2 in.) boxes
16 (2 in.) boxes
20 (2 in.) boxes
24 (2 in.) boxes
25 (2 in.) boxes
8 (100mm) boxes
9 (2 in.) boxes
Boxes (2 or 3 in.)
Dimensions (L x W x H):
415 x 140 x 168 mm
415 x 140 x 225 mm
415 x 140 x 280 mm
550 x 140 x 205 mm
550 x 140 x 225 mm
550 x 140 x 280 mm
550 x 140 x 321 mm
550 x 143 x 315 mm
679 x 140 x 225 mm
679 x 140 x 280 mm
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Dimensions (L x W x H) Unit Size
15739539 15 (2 in.) boxes 415 x 140 x 280 mm Each
15709549 20 (2 in.) boxes 679 x 140 x 225 mm Each
15719539 9 (2 in.) boxes 415 x 140 x 168 mm Each
15719549 25 (2 in.) boxes 679 x 140 x 280 mm Each
15729539 12 Boxes (2 in.) 415 x 140 x 225 mm Each
15749539 Boxes (2 or 3 in.) 550 x 143 x 315 mm Each
15759539 8 (100mm) boxes 550 x 140 x 205 mm Each
15769539 12 (3in.) boxes 679 x 140 x 280 mm Each
15779539 16 (2 in.) boxes 550 x 140 x 225 mm Each
15789539 20 (2 in.) boxes 550 x 140 x 280 mm Each
15799539 24 (2 in.) boxes 550 x 140 x 321 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15739539
  • ONLINE EXCLUSIVE PRICE
    62.60 EUR
    56.12 EUR / Pack of 1
    Save 6.48 EUR
    10% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of upright ULT freezers.
  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Side opening with handles on the front for easy box retrieval.
Specifications
Holds 15 (2 in.) boxes
Material Stainless Steel
For Use With (Application) Standard 2 in. Cryoboxes
For Use With (Equipment) Upright ULT Freezer
Dimensions (L x W x H) 415 x 140 x 280 mm
Product Type Side Access Freezer Rack