Learn More
Fisherbrand™ Upright ULT Freezer Drawer Rack for Microtiter Plates
Product Code. 15719559 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
Plates with Lid (72)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
15719559 Plates with Lid (72) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15719559
  • ONLINE EXCLUSIVE PRICE
    132.00 EUR
    117.81 EUR / Pack of 1
    Save 14.19 EUR
    10% Off
View all promotions
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
Fisherbrand inventory control rack for 96-Well and 384-Well microtiter plates. Fits all standard types of upright ULT freezers.
  • 4 drawers
  • Holds 72 plates with lid or 96 plates without lid
  • Made of high quality stainless steel
Specifications
Holds Plates with Lid (72)
Material Stainless Steel
For Use With (Application) 96/384-Well Microtiter Plates
For Use With (Equipment) Upright ULT Freezer
Dimensions (L x W x H) 547 x 247 x 138mm
Product Type Drawer Rack