Learn More
SCAT™ Universal Waste Hub 'Jan'
Artikelnummer. p-9335005 Alle ansehen SCAT Europe Produkte
Ansicht ändern
Click to view available options
Color:
Black
Blue
Thread Size:
B 53
B 63
B 83
GL 45
GL 60/61
S 50
S 51
S 55
S 60/61
S 65
S 70/71
S 90
S 95
Includes:
Earthing Cable, Safety Funnel, Splash Guard, Change Indicator (310535)
Safety Funnel, 4 PFA Blind Plugs (1.6/2.3/3.2 mm OD), 3 PTFE Blind Plugs (3 x 5 to 11.5 mm ID), Exhaust Filter M, Splash Guard, Change Indicator, Mechanical Level Control, 190 mm Floater, Earthing Cable
Safety Funnel, Splash Guard, Change Indicator (310535)
Packungsgröße:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Color Thread Size Includes Packungsgröße
17152072 Black GL 45 Earthing Cable, Safety Funnel, Splash Guard, Change Indicator (310535) Each
17162072 Black GL 60/61 Earthing Cable, Safety Funnel, Splash Guard, Change Indicator (310535) Each
17099703 Blue B 53 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17029713 Blue B 63 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17059713 Blue B 83 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17069703 Blue GL 45 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17079703 Blue S 50 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17089703 Blue S 51 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17009713 Blue S 55 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17039713 Blue S 65 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17069713 Blue S 90 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17079713 Blue S 95 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17019713 Blue S 60/61 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17049713 Blue S 70/71 Safety Funnel, Splash Guard, Change Indicator (310535) Each
17878743 Blue S 60/61 Safety Funnel, 4 PFA Blind Plugs (1.6/2.3/3.2 mm OD), 3 PTFE Blind Plugs (3 x 5 to 11.5 mm ID), Exhaust Filter M, Splash Guard, Change Indicator, Mechanical Level Control, 190 mm Floater, Earthing Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17152072 Lieferant SCAT™ Lieferanten-Nr. 420045
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The innovative Universal Waste Hub 'Jan' is a clever, all-in-one solution for liquid waste. SCAT™ offers a highly competitive number of disposal solutions around HPLC systems.
The unique multi-talent SCAT™ Universal Waste Hub 'Jan' for liquid waste disposal is a new development from SCAT™ Europe, a leading premium manufacturer for lab safety. No other product on the laboratory market for disposal solutions combines more innovative options for individual adaptation to changing disposal requirements around HPLC systems. A high number of clever details guarantees maximum safety compared to conventional solutions.
The material of the hopper is made of high quality HE-HD and is designed for long lasting use and resistant to all common chemicals used in labs. The hinge on the funnel lid is made of stainless steel and has optimal protection against corrosion. The satellite itself is made of PTFE material and ensures high stability of the entire housing. The additional dirt-sieve catches magnetic stirrers, as well as larger dirt particles. Laboratory bottles may easily be placed upon it, for rinsing purposes. The sieve can be removed for cleaning. The mesh size has been optimized and ensures for a swift throughflow of solvent. The tab with grip on the funnel lid ensures easy and convenient handling. The click closure with integrated sealing ring, prevent air pollution by uncontrolled escaping vapors. To avoid overpressure in the disposal container, the SCAT™ Universal Waste Hub 'Jan' has a connection for an original SCAT™ exhaust air filter, which is already included in the delivery. For explosive environments, two electrically conductive versions of the SCAT™ Universal Waste Hub are available that prevent sparking by using a ground cable.
All connections on the satellite are precisely processed and arranged in a very easy-to-use manner. The satellite can be securely screwed onto the disposal container and at the same time remains variably rotatable. The satellite remains flexibly usable and prevents the connected hoses from twisting. With an availability of 10 different thread sizes, all types of waste containers can be connected. Even very rare thread sizes can be connected by using optionally available adapters. The design of the Waste Hub is modern and equally appealing. The shapes of the housing and the components are ergonomically designed and have a high usability. The huge range of accessories covers all eventualities and guarantees always the right configuration of the hub.
  • Includes 1 Exhaust Filter (Size M, Polypropylene), 3 Tubes Connector (5 to 11.5 mm OD, Thread NPT 1/8 in., Polypropylene), 3 Blind Plugs (Thread NPT 1/8 in., PTFE), 1 Blind Plug (Thread GL 14, PTFE), 4 Blind Plugs (Thread UNF 1/4 in., PFA), 4 Fittings (3.2 mm ID, Thread UNF 1/4 in., PFA), 4 Fittings (2.3 mm ID, Thread UNF 1/4 in., PFA), 4 Fittings (1.6 mm ID, Thread UNF 1/4 in., PFA)
Spezifikation
For Use With (Application) Liquid Waste Disposal
For Use With (Equipment) HPLC and UHPLC Systems
Type Disposal Unit
Solvent Channels 7
Material HDPE-EC, PTFE, PFA
Includes Earthing Cable, Safety Funnel, Splash Guard, Change Indicator (310535)
Quantity 1 Set
Color Black
Thread Size GL 45