Learn More
Fisherbrand™ Universal pH Indicator Paper
Product Code. 10455532 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
1 to 14
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10455532 1 to 14 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10455532
  • ONLINE EXCLUSIVE PRICE
    7.15 EUR
    6.41 EUR / Pack of 200
    Save 0.74 EUR
    10% Off
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
  • Filter paper soaked with one or several colored indicators of pH
  • Economic range of pH 0 to 14
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
pH Range 1 to 14
Format Box
Quantity 200 Box
Type Grade 911