Learn More
Heidolph™ Perforated Platform 1000
Product Code. 10322702 Shop All Heidolph Instruments Products
Change view
Click to view available options
Speed Range:
0 to 1100 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
10322702 0 to 1100 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10322702 Supplier Heidolph™ Supplier No. 5495910000
Estimated Shipment: 07-01-2027
For use with Vibramax 100 and Rotamax 120
Universal perforated platform allows for individual arrangement of vessels
TRUSTED_SUSTAINABILITY
Specifications
Load Bearing Capacity (Metric) 2 kg
Speed Range 0 to 1100 rpm
Product Type Perforated Platform
Dimensions (L x W) 22 x 22 cm