Learn More
Hughes Unheated Mobile Safety Shower with Eye/Face Wash
Product Code. 17722358
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17722358 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17722358 Supplier Hughes Supplier No. HSHS040KGV
  • ONLINE EXCLUSIVE PRICE
    6575.31 EUR
    4916.87 EUR / Pack of 1
    Save 1658.44 EUR
    25% Off
 Restricted Item
Estimated Shipment: 14-12-2026
The 114 liter capacity unheated mobile self-contained emergency safety shower with eye/face wash unit. Incorporating a polypropylene lined cylinder, a stainless steel frame and large pneumatic tires, making it easy to maneuver by one person.
  • The 114 liter capacity unheated mobile self-contained emergency safety shower with eye/face wash unit
  • Incorporating a polypropylene lined cylinder, a stainless steel frame and large pneumatic tyres, making it easy to manoeuvre by one person
  • For long distances or uneven surfaces an optional 'A' frame with towing hitch can be fitted
  • In addition, the wheel spacing can be altered allowing the unit to fit through standard door openings
Specifications
Capacity (Metric) 114 L
Color White
Product Type Safety Shower