Learn More
Haldenwanger™ Unglazed Porcelain Pestle
Product Code. 12313338
Change view
Click to view available options
Length (Metric):
115mm
125mm
135mm
150mm
175mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12313338 125mm Each
10656405 115mm Each
10042964 150mm Each
10287274 135mm Each
12343338 175mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12313338 Supplier Haldenwanger™ Supplier No. L 56/0A/RAUH
  • ONLINE EXCLUSIVE PRICE
    15.60 EUR
    13.56 EUR / Pack of 1
    Save 2.04 EUR
    13% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
Use these pestles for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Pestles are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • Grinding surfaces are unglazed
  • Complies with DIN VDE 0335, type C 100
Specifications
Product Type Pestle
Material Porcelain
Length (Metric) 125mm
Finish Unglazed