Learn More
Köttermann Underbench Safety Cabinet
This Underbench Safety Cabinet is ideal for flammable hazardous substances, complying with fire resistance class standards (Type 90).
Brand:  Köttermann 371-00032
Additional Details : Weight : 259.00000kg
Product Code. 17392129
  • 4070.40 EUR / Each
Estimated Shipment: 16-09-2024
Description
  • Low design to fit under laboratory bench
  • Fire resistant (Type 90)
  • Features two pull-out doors
Specifications
Specifications
570 x 1400 x 720 mm
Drawer
2