Learn More
Invitrogen™ UltraPure™ Cesium Chloride, Optical Grade
Product Code. 10073482
Change view
Click to view available options
Quantity:
1 kg, 1kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10073482 1 kg, 1kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10073482 Supplier Invitrogen™ Supplier No. 15507023
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Edge
Add to basket
UltraPure™ Cesium Chloride, Optical Grade is specifically prepared for use in density gradients.
UltraPureTM Cesium Chloride, Optical Grade is specifically prepared for use in density gradients. The highest grade commercially available, UltraPureTM Cesium Chloride, Optical Grade can be used to prepare plasmids or to isolate phage, virus particles, or cellular RNA.
UltraPure™ Cesium Chloride, Optical Grade is specifically prepared for use in density gradients. The highest grade commercially available, UltraPure™ Cesium Chloride, Optical Grade can be used to prepare plasmids or to isolate phage, virus particles, or cellular RNA.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cesium Chloride
Content And Storage Store at room temperature.
Product Line UltraPure
Quantity 1 kg, 1kg
Shipping Condition Room Temperature
Form Powder