Learn More
IKA™ T 50 Digital ULTRA-TURRAX™ Disperser
GREENER_CHOICE
Product Code. 13306309 Shop All IKA Products
Change view
Click to view available options
Speed:
600 to 10,000 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed unitSize
13306309 600 to 10,000 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13306309 Supplier IKA™ Supplier No. 0003787000
Only null left
Add to basket
Add to basket
Offering a wide speed range from 500 to 10,000rpm that enables users to work at high circumferential speeds even with small rotor diameters.
  • Powerful dispersing instrument for volumes from 0.25 to 30L (H2O) with digital speed display
  • Broad choice of dispersing elements guarantees a wide range of applications
  • Reproducible operations due to constant speed even with changes in viscosities
  • Electronic speed control
  • Electronic overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Smooth start
  • Error code display
TRUSTED_SUSTAINABILITY
Specifications
Product Type Digital Disperser
Speed 600 to 10,000 rpm
Voltage 220/240 V
Electrical Requirements 220/240 V, 50/60 Hz, 1100 W
Hertz 50/60 Hz
Wattage 1100 W
Volume Range (Metric) 250 mL to 30 L
Depth (Metric) 139 mm
Height (Metric) 355 mm
Width (Metric) 115 mm
Display LED
For Use With (Application) Dispersing at high circumferential speeds with small rotor diameters
Model T 50 ULTRA-TURRAX™
Packaging Quantity 1 Ea.
Viscosity 5000 mPas
Weight (Metric) 5.76 kg