Learn More
IKA™ T 10 Basic ULTRA-TURRAX™ Disperser
Product Code. 13316309 Shop All IKA Products
Change view
Click to view available options
Speed:
8000 to 30,000 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Unit Size
13316309 8000 to 30,000 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13316309 Supplier IKA™ Supplier No. 0003737000
  • ONLINE EXCLUSIVE PRICE
    1615.00 EUR
    1525.97 EUR / Pack of 1
    Save 89.03 EUR
    5% Off
Estimated Shipment: 11-12-2026
A small dispersing instrument for volumes from 0.5 to 100mL (H2O) that offers a wide speed range, enabling users to work at high circumferential speeds even with small rotor diameters.
  • Speed range of 8000 to 30,000rpm
  • Broad choice of dispersing elements guarantees a wide range of applications
  • Quick-connect coupling for changing dispersing elements easily
  • Ideal for manual operation due to its light weight and ergonomic design
  • Extremely mobile due to direct line power (no transformer required)
  • Stainless steel dispersing elements can be cleaned quickly and easily as they can be dismounted without tools
  • Plastic disposable dispersing elements are available in two sizes, particularly suitable for PCR analysis
  • High performance drive ensures immense speed stability
  • Quiet operation
TRUSTED_SUSTAINABILITY
Specifications
Speed 8000 to 30,000 rpm
Voltage 230 V
Electrical Requirements 230 V, 50/60 Hz, 125 W
Wattage 125 W
Volume Range (Metric) 0.5 to 100 mL
Depth (Metric) 66 mm
Height (Metric) 178 mm
Width (Metric) 56 mm
For Use With (Application) PCR analysis
Model T 10 ULTRA-TURRAX™
Packaging Quantity 1 Ea.
Viscosity 5000 mPas
Product Type Basic Disperser
Frequency 50/60 Hz