Learn More
B Braun™ Sterican™ Single-use Needles
Description
- Needle made of stainless chromium-nickel steel with extremely smooth surface and light silicone coating for minimal pain upon puncture.
- Transparent Luer Lock plastic hub made of polypropylene
- Colour-coded hub identifies the outer diameter of the needle, conforms to ISO 6009
- Latex-free/PVC-free
Specifications
Specifications
| Sterility | Sterile |
| Cannula Material | Stainless Chromium Nickel Steel |
| Certifications/Compliance | EN ISO 7864 and DIN 13097 |
| Closure Type | Luer Lock Hub |
| Color | Green |
| Diameter (Metric) Outside | 0.80 mm |
| For Use With (Application) | Withdrawal and flushing |
| Gauge | 21 G |
| Hub Material | Polypropylene |
| Latex Free | Yes |
| Show More |