Learn More
Thermo Scientific™ Ultra Pure Water Distributors
Product Code. 15590226 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
1 inlet, 2 outlets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15590226 1 inlet, 2 outlets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15590226 Supplier Thermo Scientific™ Supplier No. 50155463
  • ONLINE EXCLUSIVE PRICE
    233.00 EUR
    215.00 EUR / Pack of 1
    Save 18.00 EUR
    7% Off
 Restricted Item
Estimated Shipment: 28-01-2027
Use Thermo Scientific™ Ultra Pure Water Distributors with all SDI systems.
Use Thermo Scientific™ Ultra Pure Water Distributors with all SDI systems.
  • Plastic
  • Connectors allow connection of shut-off taps
TRUSTED_SUSTAINABILITY
Specifications
Description 1 inlet, 2 outlets
For Use With (Equipment) SDI System
Type Ultra Pure Water Distribution Manifold
Inlet Connection NPTF (English) 1 x R 3/4 in. union nut; 2 x R3/4 in. male thread
Material PVC
Width (Metric) 280 mm
Height (Metric) 170 mm
Depth (Metric) 50 mm