Learn More
UDP-3-O[R-3-Hydroxymyristoyl]-N-acetylglucosamine Tris Salt (90%), TRC
Product Code. 30455405
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455405 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455405 Supplier Toronto Research Chemicals Supplier No. TRCU2505255MG
 Restricted Item
Estimated Shipment: 10-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material UDP-3-O-[(R)-3-Hydroxymyristoyl]-N-acetyl-D-glucosamine Tris Salt
CAS 112710-84-6
Molecular Formula C35H64N4O22P2
Quantity 5 mg
Synonym 1,3-Propanediol','2-amino-2-(hydroxymethyl)-P'-[2-(acetylamino)-2-deoxy-3-O-(3-hydroxy-1-oxotetradecyl)-α-D-glucopyranosyl] Ester','Compd. With Uridine 5'-(trihydrogen diphosphate), UDP-3-O[R-3-Hydroxymyristoyl]-GlcNAc Tris Salt
InChI Formula InChI=1S/C31H53N3O19P2.C4H11NO3/c1-3-4-5-6-7-8-9-10-11-12-19(37)15-23(39)51-28-24(32-18(2)36)30(50-20(16-35)26(28)41)52-55(46,47)53-54(44,45)48-17-21-25(40)27(42)29(49-21)34-14-13-22(38)33-31(34)43;5-4(1-6,2-7)3-8/h13-14,19-21,24-30,35,37,40-42H,3-12,15-17H2,1-2H3,(H,32,36)(H,44,45)(H,46,47)(H,33,38,43);6-8H,1-3,5H2/t19-,20-,21-,24-,25-,26-,27-,28-,29-,30-;/m1./s1
SMILES O[C@@H]1[C@H](O)[C@@H](COP(O)(OP(O[C@@H]2[C@H](NC(C)=O)[C@@H](OC(C[C@H](O)CCCCCCCCCCC)=O)[C@H](O)[C@@H](CO)O2)(O)=O)=O)O[C@H]1N(C=C3)C(NC3=O)=O.NC(CO)(CO)CO
IUPAC Name 1,3-Propanediol','2-amino-2-(hydroxymethyl)-','compd. with uridine 5′-(trihydrogen diphosphate) P′-[2-(acetylamino)-2-deoxy-3-O-(3-hydroxy-1-oxotetradecyl)-α-D-glucopyranosyl] ester (1:1)
Molecular Weight (g/mol) 954.84
Formula Weight 954.35