missing translation for 'onlineSavingsMsg'
Learn More

Udenafil, MedChemExpress

Product Code. 16493507
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493507 50 mg 50mg
16487446 100 mg 100mg
16469967 5 mg 5mg
16447488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493507 Supplier Medchem Express Supplier No. HY18253
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Udenafil (DA8159) is a potent, selective and orally active phosphodiesterase type 5 (PDE5) inhibitor. Udenafil also inhibits cGMP hydrolysis and can be used for erectile dysfunction research.

Udenafil (DA8159) is a potent, selective and orally active phosphodiesterase type 5 (PDE5) inhibitor. Udenafil also inhibits cGMP hydrolysis and can be used for erectile dysfunction research.

Specifications

Chemical Name or Material Udenafil
CAS 268203-93-6
Color White
Purity Grade Notes Research
Molecular Formula C25H36N6O4S
Quantity 50 mg
Synonym DA8159
Solubility Information DMSO : ≥ 33 mg/mL (63.87 mM)
SMILES O=S(C1=CC=C(OCCC)C(C2=NC3=C(N(C)N=C3CCC)C(N2)=O)=C1)(NCCC4N(C)CCC4)=O
Molecular Weight (g/mol) 516.66
Formula Weight 516.66
Percent Purity 99.86%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.