missing translation for 'onlineSavingsMsg'
Learn More

UBE2N/Ubc13 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18513681 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18513681 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18513681 Supplier Novus Biologicals Supplier No. NBP298828AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBE2N/Ubc13 Polyclonal antibody specifically detects UBE2N/Ubc13 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2N/Ubc13
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias bendless-like ubiquitin conjugating enzyme, Bendless-like ubiquitin-conjugating enzyme, BLU, EC 6.3.2.19, MGC131857, MGC8489, UBC13, UbcH-ben, Ubiquitin carrier protein N, ubiquitin-conjugating enzyme E2 N, ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13), ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast), Ubiquitin-protein ligase N, yeast UBC13 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human UBE2N/Ubc13.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7334
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.